Jerricans, HDPE, with screw cap
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2828 2825 2826 2829
SEMA2828EA 78.7 EUR
SEMA2828 SEMA2825 SEMA2826 SEMA2829
Jerricans, HDPE, with screw cap
Flessen Jerrycans
HDPE, translucent.
  • Good chemical resistance
  • Good temperature rsistance
  • Stackable
  • UN approved