Porcine serum
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S2400-100 S2400-500 S240H-500 S240H-100
S2400-100EA 26.4 EUR
S2400-100 S2400-500 S240H-500 S240H-100
Porcine serum
Serum
Porcine serum in lymphocyte culture for chromosomal studies has been utilized and found to be adequate. Moreover, although not commonly used, it is said that the constituents of porcine and human blood are nearly the same. It has also been suggested that porcine serum could be used in setting up peripheral lymphocyte culture for karyotyping.