Guinea pig serum
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S2450-100 S2450-500 S2450-050
S2450-100EA 381 EUR
S2450-100 S2450-500 S2450-050
Guinea pig serum
Serum
Guinea pig serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Guinea pig serum is used as a blocking agent in immunoassays and controls.