Analytical HPLC columns, XTerra® MS C18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
186000440 186000878 186000422 186000474 186000494 186000660 186000632 186000432 186000400
WATE186000440EA 0 EUR
WATE186000440 WATE186000878 WATE186000422 WATE186000474 WATE186000494 WATE186000660 WATE186000632 WATE186000432 WATE186000400
Analytical HPLC columns, XTerra® MS C18
Kolommen voor chromatografie
XTerra® MS C18 columns are general purpose, hybrid-based, reversed phase C18 columns that are designed to be compatible with mass spectrometry applications. These first generation columns provide superior pH stability compared to a silica based column.