Transport tubes, with stopper
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1696 1698 1699
212-3232EA 58.2 EUR
212-3232 SEMA1698 SEMA1699
Transport tubes, with stopper
Buisjes Reageerbuisjes
PS, clear, with white push-in stopper.
  • Flat bottom
  • Good thermal resistance
  • Good chemical resistance
Universal tubes for a wide range of applications.