Analytical HPLC columns, XTerra® RP18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
186000434 186000662 186000496 186000492 186000442 186000410
WATE186000434EA 0 EUR
WATE186000434 WATE186000662 WATE186000496 WATE186000492 WATE186000442 WATE186000410
Analytical HPLC columns, XTerra® RP18
Kolommen voor chromatografie
XTerra® RP18 columns are general purpose, hybrid-based, reversed phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.