Reversed phase HPLC columns, Atlantis® Silica HILIC
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
186002016 186002012
WATE186002016EA 0 EUR
WATE186002016 WATE186002012
Reversed phase HPLC columns, Atlantis® Silica HILIC
Kolommen voor chromatografie
Atlantis HILIC silica columns are used in combination with high organic mobile phases (>80% acetonitrile) to provide retention of analytes that are simply too polar to be retained by reverse-phase chromatography.