Reversed phase HPLC columns, µBondapak® C18
Leverancier: WATERS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WAT027324 WAT086684 WAT088070 186000925 WAT044480
WATEWAT027324EA 0 EUR
WATEWAT027324 WATEWAT086684 WATEWAT088070 WATE186000925 WATEWAT044480
Reversed phase HPLC columns, µBondapak® C18
Kolommen voor chromatografie
µBondapak® C18 columns are general purpose, silica-based, reversed phase C18 columns that are based on 10 µm particle technology.