Tubing, silicone
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4348 4350 4353 4349
SEMA4348EA 6.5 EUR
SEMA4348 SEMA4350 SEMA4353 SEMA4349
Tubing, silicone
Slangen Transfer Tubing
Silicone, translucent.
  • Non toxic
  • Excellent chemical resistance
  • Very flexible
Temperature resistance: −50 to +180 °C
Hardness (Shore durometer A): 60