Safety disposal unit (funnel-model) with drain tray
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4500 4501 4502 4503
SEMA4500EA 744 EUR
SEMA4500 SEMA4501 SEMA4502 SEMA4503
Safety disposal unit (funnel-model) with drain tray
Veiligheidscontainers
Safety disposal canister in a drain tray made of polypropylene (PP) for the disposal of hazardous waste.
  • With a screw-on funnel
  • Inward sieve and level indicator to avoid overfilling
  • Large opening, self closing lid to prevent odour and corrosion in the room