Jars, wide mouth, square, with screw cap
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3743 3744 4457
SEMA3743EA 9.6 EUR
SEMA3743 SEMA3744 SEMA4457
Jars, wide mouth, square, with screw cap
Potjes Potjes voor algemeen gebruik
Jars, wide mouth, made of PET, crystal clear, with white screw cap.
  • Perfectly glassy and prominently present
  • Elegant, angular shape with white screw cap
Suitable for powders, granules, pastes or foodstuffs. Good chemical resistance to aqueous solutions of mineral acids, salts, soaps, oils, alcohols and aliphatic hydrocarbons. Suitable for alkaline solutions, solvents or strong acids.