SPE microelution plates, SOLAµ™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60209-001 60209-002 60209-003 60209-004 60209-005
549-0879EA 329.8 EUR
549-0879 549-0880 549-0881 549-0882 549-0883
SPE microelution plates, SOLAµ™
Microplaten Platen voor vastefase-extractie
SOLA™μ 96 well SPE micro elution plates are specifically designed to meet the increasingly demanding challenges found within bioanalysis. The micro elution format is uniquely positioned to deliver on bioanalytical needs, but limitations with existing
technological design has prevented significant uptake. The SOLA™μ product eliminates these issues by utilising SOLA technology, which removes the need for frits to retain the sorbent material and, in combination with a macro porous structure, allows for a robust, easily controllable platform for reproducible processing of bioanalytical samples.