Microcentrifuge tubes, 0,6/1,6/2,0 ml
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
710200 710201 710202 710203 710204 710205 710206 710208 710210 710220 710290 710291
BZYM710200EA 0 EUR
BZYM710200 BZYM710201 BZYM710202 BZYM710203 BZYM710204 BZYM710205 BZYM710206 BZYM710208 BZYM710210 BZYM710220 BZYM710290 BZYM710291
Microcentrifuge tubes, 0,6/1,6/2,0 ml
Buisjes Microcentrifugebuisjes
PP, translucent.
  • DNase-, RNase-free
Graduated, with safelock cap.