Strips of 8 PCR caps, 0,2 ml, SoftStrips®
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
710980 710981 710982 710983 710984 710986 710988 710990 710991 710992 710993 710994 710996 710998 710971 710970
BZYM710980EA 0 EUR
BZYM710980 BZYM710981 BZYM710982 BZYM710983 BZYM710984 BZYM710986 BZYM710988 BZYM710990 BZYM710991 BZYM710992 BZYM710993 BZYM710994 BZYM710996 BZYM710998 BZYM710971 BZYM710970
Strips of 8 PCR caps, 0,2 ml, SoftStrips®
Buisjes PCR-buisjes
Available as single cap strip or attached cap strips.
  • Optical clear
  • High profile
  • Free from DNA, DNase and RNase