Lab markers, Teeny Writers
Leverancier: DIVERSIFIED BIOTECH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TNY-WTR TNY-TWIN
DIVETNY-WTREA 16.5 EUR
DIVETNY-WTR DIVETNY-TWIN
Lab markers, Teeny Writers
Pennen Stiften voor laboratoria
A retractable Sharpie® pen with an extra fine tip. Recommended for writing on all our labels for samples that will be stored at room temperature and not exposed to chemicals and/or caustic agents.