Clamps
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5409 5407 5406 5408
BOCH5409EA 56.1 EUR
BOCH5409 BOCH5407 BOCH5406 BOCH5408
Clamps
Klemmen Klemmen voor algemeen gebruik
Retort clamps made of stainless steel.
  • Jaws are covered with a layer of cork to protect clamped items
  • Angled jaws
Certificaten: DIN 12894