Storage Boxes for Vials
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
702514 702515 702516 702517 702518
MANA702514EA 15.7 EUR
MANA702514 MANA702515 MANA702516 MANA702517 MANA702518
Storage Boxes for Vials
Dozen Dozen voor cryobuisjes
Boxes are made from blue plastic with integrated or removable divider and transparent lid.
  • Allow a secure transportation of sample vials
  • Safe standing position in dividers designed for the respective diameter
  • Available for all 1,5 ml vials (standard volume), for crimp and screw neck vials N 13 and for headspace vials with screw neck N 18 or crimp neck N 20, respectively
  • Also available for EPA screw neck vials N 24 as well as for storage vials N 15
Ideal for space-saving storage in fridges and freezers, since the transparent lid prevents condensations on the closures and thus avoids a possible contamination in the cooling unit.