Double bossheads
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5337 5365
241-3177EA 23.3 EUR
241-3177 BOCH5365
Double bossheads
Klemmen Statiefkoppelingen
Double bossheads, either made of aluminium or malleable cast iron, chromium plated.
  • For rods up to 16,5 mm
  • Angle: 90°
  • With safety screw type 'heart' (M8)
Certificaten: DIN 12895