Disposable cuvettes
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3549 3892 2531
216-1267EA 75.8 EUR
216-1267 612-0610 SEMA2531
Disposable cuvettes
Cuvetten Cuvetten voor spectrofotometers
Disposable cuvettes for photometric measurements in the visible range (350 to 800 nm).