Mengmaatcilinders, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
246180856 246181458 246181758 246182454 246183656 246184455 246185451 246186353
215-3671EA 50.3 EUR
215-3671 215-3672 215-3673 215-3674 215-3675 215-3676 215-3677 215-3678
Mengmaatcilinders, DURAN®
Cilinders Maatcilinders
DURAN® borosilicaatglas, met PE-stop, klasse A.
  • Blue graduation, with main points ring graduation
  • With certificate of conformity 'DE-M'
With ergonomic PE stopper and hexagonal base.
With printed retrace code (batch identification).
Order Now
Specificaties
Environmentally Preferable