Cryoboxes
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5150021 5150022 5150023 5150002 5150003 5150012 5150013 5150014 5150011
RATI5150021EA 70.1 EUR
RATI5150021 RATI5150022 RATI5150023 RATI5150002 RATI5150003 RATI5150012 RATI5150013 RATI5150014 RATI5150011
Cryoboxes
Dozen Dozen voor cryobuisjes
PC, coloured, with clear lid.
  • Temperature-resistant from −190 to 121 °C
  • Drain bores at the bottom for dew liquids
  • By stacking corners the boxes are suitable for safe transport
With slip lid and variable grid insert.