Multipurpose container, round, with snap-on lid
Leverancier: hünersdorff GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
913100 913200
216-3740EA 0 EUR
216-3740 216-3741
Multipurpose container, round, with snap-on lid
Monsternamepotjes Containers voor algemeen gebruik
HDPE, translucent.
  • Resistant to acids and chemicals
  • With tightly closing lid
  • Deep freezable
  • Stackable
For storing and transporting solid and liquid materials, samples and general labware.