Heating tapes, HST
Leverancier: HORST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
025207 025201 025202 025203 025204 025205 025206
HORS025207EA 673 EUR
HORS025207 HORS025201 HORS025202 HORS025203 HORS025204 HORS025205 HORS025206
Heating tapes, HST
Verwarmers
These slim heating tapes consist of two PTFE insulated heating conductors with earthed protection braid which have a common coating made of metal cloth.