Roll mop, PolyTack®
Leverancier: CONNECTICUT CLEANROOM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PH-18 PR-18F PR-9F PR-18
CONNPH-18EA 267 EUR
CONNPH-18 CONNPR-18F CONNPR-9F CONNPR-18
Roll mop, PolyTack®
Moppen
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
  • Easy to use hand or pole attachments
  • Plastic inner core insures cleanliness
  • Anti-microbial agent providing protection against bacterial growth