ESD occupational shoes, slip-on, Zwiesel
Leverancier: SCHURR SCHUHVERTRIEB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11118.01.51/43 11118.01.51/44 11118.01.51/47 11118.01.51/45
SCRR11118.01.51/43EA 79.3 EUR
SCRR11118.01.51/43 SCRR11118.01.51/44 SCRR11118.01.51/47 SCRR11118.01.51/45
ESD occupational shoes, slip-on, Zwiesel
Schoenen en laarzen
These ESD shoes are made from non perforated microfibre upper with a comfortable anti-slip PU sole. They are suitable for kitchen applications.
  • DRY TECH textile lining
  • Interchangeable anti-static PUR foam insole
  • Good profiled sole with raised heel
Colour: White
Certificaten: EN 61340 (ESD); EN ISO 20347:2004