SPE cartridges and columns, Supra-Clean®, C18
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9306448 N9306422 N9306430 N9306438 N9306462 N9306476 N9306478 N9306479 N9306523 N9306589
PERCN9306448EA 128 EUR
PERCN9306448 PERCN9306422 PERCN9306430 PERCN9306438 PERCN9306462 PERCN9306476 PERCN9306478 PERCN9306479 PERCN9306523 PERCN9306589
SPE cartridges and columns, Supra-Clean®, C18
Kolommen voor chromatografie
  • Pure spherical silica
  • Pore size 60 to 120 Å
  • pH range 2 to 8