SPE columns, Supra-Poly® XWP
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9306418 N9306427 N9306504 N9306505 N9306506 N9306507 N9306510
PERCN9306418EA 475 EUR
PERCN9306418 PERCN9306427 PERCN9306504 PERCN9306505 PERCN9306506 PERCN9306507 PERCN9306510
SPE columns, Supra-Poly® XWP
Kolommen voor chromatografie
SPE Supra-Poly® extreme capacity wide pore (XWP) are polymer based columns, polystyrene-divinylbenzene (PSDVB). Ideal for extracting polar and non-polar compounds when analysing drugs and drug metabolites in biological fluids.