SPE columns, Supra-Clean® SI-S (silica)
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
N9306444 N9306402 N9306404 N9306466 N9306532 N9306533 N9306534 N9306535
PERCN9306444EA 174 EUR
PERCN9306444 PERCN9306402 PERCN9306404 PERCN9306466 PERCN9306532 PERCN9306533 PERCN9306534 PERCN9306535
SPE columns, Supra-Clean® SI-S (silica)
Kolommen voor chromatografie
SPE Supra-Clean® SI-S are spherical silica-based columns. Ideal for extracting polar compounds when analysing aldehydes, amines, pesticides, herbicides, carotenoids, fat soluble vitamins, baflatoxins, fatty acids, and phospholipids.