Occupational shoes, clogs, Super Birki WL8011
Leverancier: LIEFERSERVICE UND LOGISTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
068021/43 068011/42 068011/43 068011/44 068011/40 068011/46 068011/38 068651/40 068651/43 068011/37 068011/39 068071/46
LIEF068021/43EA 87 EUR
LIEF068021/43 LIEF068011/42 LIEF068011/43 LIEF068011/44 LIEF068011/40 LIEF068011/46 LIEF068011/38 LIEF068651/40 LIEF068651/43 LIEF068011/37 LIEF068011/39 LIEF068071/46
Occupational shoes, clogs, Super Birki WL8011
Schoenen en laarzen
The Super-Birki anti-static clog models are fitted with replaceable anti-static integrated insoles. Birki anti-static model has an integrated insole made of a single piece of flexible anti-static blended foam.
  • Anti-static and slip-resistant
  • Alkali, oil and grease resistant
  • Washable at 60 °C
EN ISO 20347
Certificaten: EN ISO 20347