Wipes, pre-saturated
Leverancier: ESSENTRA POROUS TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6-C86-EFP 6-C43
EPTA6-C86-EFPEA 579 EUR
EPTA6-C86-EFP EPTA6-C43
Wipes, pre-saturated
Schoonmaakdoekjes Doekjes en handdoeken voor eenmalig gebruik Vochtige doekjes
These heavyweight polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
  • Pre-wetted with high concentration of alcohol
  • Individually packed: Easy tear, single use foil package
  • Dissolves most residues safely and effectively
  • Excellent as a degreaser
  • Made in the USA
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904