Outdoor sealing mat, for drains
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9901 9902 9903 9904 13098
MSGA9901EA 418 EUR
MSGA9901 MSGA9902 MSGA9903 MSGA9904 MSGA13098
Outdoor sealing mat, for drains
Vloermatten Permanente vloermatten
Sealing mats made from a dense and flexible material prevent fast and efficient penetration of water-polluting liquids into the ground or drains.
  • Rapid protection against spilled liquids
  • Resistant to hydrocarbon and most chemicals
  • Reusable, can be washed of with soap and water
Colour: White/yellow