Mat frames for PurusMat
Leverancier: PURUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PF284701W00 PF384701W00 PF204701W00
PURUPF284701W00EA 432 EUR
PURUPF284701W00 PURUPF384701W00 PURUPF204701W00
Mat frames for PurusMat
Vloermatten Matframes
These frames are designed to be used with the Purus mats to create a system that is ideal for any critical area that does not allow direct adhesion to the flooring surface. The frames are constructed of a hard, rigid, PS base material combined with a non skid backing that holds the mats in place on almost any surface.
  • Protect floors from adhesive residue from the purus mats
  • Reduces the risk of slips and falls
  • Non-slip parts made from polymer coated polyester textile
  • Naturally antimicrobial, mould and mildew resistant