Dropping funnels, cylindrical, graduated, with pressure balance
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1582-97-130 1582-97-102 1582-97-120
NEUB1582-97-130EA 396 EUR
NEUB1582-97-130 NEUB1582-97-102 NEUB1582-97-120
Dropping funnels, cylindrical, graduated, with pressure balance
Trechters Druppeltrechters
Borosilicate glass 3.3, clear, with solid glass key.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Standard ground joint cone and socket, with divisions for approximate volume reading.