Washable cleanroom floor mats, CleanZone
Leverancier: DYCEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CZ01/12/SL CZ01/20/C0 CZ01/20/MN
DYCECZ01/12/SLEA 2220 EUR
DYCECZ01/12/SL DYCECZ01/20/C0 DYCECZ01/20/MN
Washable cleanroom floor mats, CleanZone
Vloermatten Permanente vloermatten
CleanZone is a high performance contamination control flooring suitable for light wheeled traffic and heavier pedestrian flows. It is washable and a most effective way of controlling all forms of foot, wheel and air borne contamination.
  • Flexible to be installed at different sized and shaped areas
  • Prevents over 99% of contamination from entering critical area
  • Reduces floor and air particle and microbial counts
  • Improved yield, cost savings and profitability
  • Available as flame retardant, coving and inlaid options
Colour: Light or dark blue, dark grey