Tissue adhesives, Histoacryl®
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9381104 1050044
BRAU9381104EA 701 EUR
BRAU9381104 BRAU1050044
Tissue adhesives, Histoacryl®
Eerste-hulpbenodigdheden Wondverbanden
The tissue adhesive Histoacryl® consists of monomeric n-butyl-2-cyanoacrylate, which polymerises quickly in connection with tissue fluid. The adhesive can be employed for non bleeding and for bleeding varices.
  • Wound closure in one minute
  • Anti-bacterial barrier protects the wound
  • Can be stored at room temperature
  • Excellent cosmetic results
  • Easy-to-use procedure
Ideal for the closure of smooth and fresh skin wounds, for non traumatic mesh fixation in hernia repair and for the closure of skin in endoscopic incisions. Combined therapy with Histoacryl® and conventional sclerosation therapy is possible.