Disinfectant, ClearKlens IPA 70%, sterile
Leverancier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
141-0889EA 252 EUR
141-0889 141-1407 DIVY7516050 DIVY7514372 141-1422 DIVY101104902
Disinfectant, ClearKlens IPA 70%, sterile
Reinigingsmiddelen Desinfectiemiddelen
Disinfectant containing 70% isopropyl alcohol, 30% water for injection, for use on contact surfaces, gloved hands, instruments and utensils.
  • Sterile disinfectant with 70% isopropanol / 30% WFI
  • Ready to use product for the disinfection of work surfaces and process equipment in cleanrooms
  • 0,2 μm filtered, gamma irradiated/aseptically filled and double-bagged
  • ClearKlens® IPA 900 ml trigger spray is an anti-suck back, 'bag in bottle' delivery system to eliminate product contamination during use
  • ClearKlens® IPA 1 L bottle/5 L canister is used for mopping and wiping applications - the 1 L bottle can be used with the dedicated touchless dispenser for glove disinfection
  • BPR approved pack and formulation
Standards: ClearKlens® IPA has been extensively tested using EN 1276, EN 1650 and EN 13697. These have shown ClearKlens® IPA to have a broad spectrum of activity against both gram-positive/gram-negative bacteria and yeasts. Tests according to EN 14476 have shown a biocidal efficacy against enveloped viruses (modified vaccinia virus Ankara/MVA) like SARS-CoV-2. Full test data is available as part of the technical document package.
Opgelet: Use biocides safely. Always read the label and product information before use.