Adhesive strips
Leverancier: DYCEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
NS05PA1/1 NS05/ST/1
DYCENS05PA1/1EA 21.3 EUR
DYCENS05PA1/1 DYCENS05/ST/1
Adhesive strips
Etiketten
Self-adhesive strips and panels are manufactured from Dycem multipurpose reel material with an adhesive backing added to secure more permanently and give an enhanced built up grip.
  • Versatile and easy to wrap around a variety of items
  • Panels used to line a splint, increase friction and prevent the splint from slipping and shifting
  • Strips are also available in comfort material, providing a thicker and cushioned non-slip grip