Veterinary blood collection tubes with clot activator, vacuum, Vacutest®
Leverancier: KIMA SAS DI CHIARIN RENZO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11010V 11011V
720-2732EA 2110 EUR
720-2732 720-2733
Veterinary blood collection tubes with clot activator, vacuum, Vacutest®
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The test tubes are made from polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use (40×20 mm), printed in three colours.
  • Available in different sizes
  • 18 months shelf-life
The closure system of the test tubes is composed of a pierceable butyl rubber stopper and a polyethylene safety cap. The stopper has a hollow in the piercing spot to protect operator from possible contact. The safety cap protecting against the aerosol and splashes prevents the operator from coming into contact with the pierceable rubber stopper and allows for easy recapping of the test tube.
Certificaten: Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes