Safety waste caps and ventilation kit, SymLine®
Leverancier: S.C.A.T.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
106480 106478 106484 106507 106527 106522 306482 306509 306581 106718 106719 106720
SCTV106480EA 296.46 EUR
SCTV106480 SCTV106478 SCTV106484 SCTV106507 SCTV106527 SCTV106522 SCTV306482 SCTV306509 SCTV306581 SCTV106718 SCTV106719 SCTV106720
Safety waste caps and ventilation kit, SymLine®
Sluitingen Veiligheidsdoppen en veiligheidsafvaldoppen
Safety waste caps safely connect pipework to the waste container. The integrated exhaust filter blocks solvent vapours, for the protection of health and the environment.
  • Caps are available with different thread sizes for attachment to the pipe system
  • Caps are available with different numbers of hose connections for disposal of liquid waste
  • By using a 4-in-1 collector, up to 4 safety waste caps can be connected simultaneously to only one connection point on the pipe system
  • Ventilation tube, length 1500 mm, allows connection to lab exhaust system
  • Adapter (Y-