Occupational shoes, slip-on, Alpro®, A 640
Leverancier: LIEFERSERVICE UND LOGISTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
020252/47 020272/46 020292/46 020292/45 020292/44 020292/43 020292/42 020292/41 020292/40 020292/39 020292/38 020292/37 020262/45 020272/47 020292/47 020272/45 020272/44 020272/43 020272/42 020272/41 020272/40 020272/39 020272/38 020272/37 020272/36 020292/36 020252/45 020262/44 020262/43 020262/42 020262/41 020262/40 020262/39 020262/38 020262/37 020262/36 020262/46 020252/46 020262/47 020252/44 020252/43 020252/42 020252/41 020252/40 020252/39 020252/38 020252/37 020252/36
LIEF020252/47EA 69.3 EUR
LIEF020252/47 LIEF020272/46 LIEF020292/46 LIEF020292/45 LIEF020292/44 LIEF020292/43 LIEF020292/42 LIEF020292/41 LIEF020292/40 LIEF020292/39 LIEF020292/38 LIEF020292/37 LIEF020262/45 LIEF020272/47 LIEF020292/47 LIEF020272/45 LIEF020272/44 LIEF020272/43 LIEF020272/42 LIEF020272/41 LIEF020272/40 LIEF020272/39 LIEF020272/38 LIEF020272/37 LIEF020272/36 LIEF020292/36 LIEF020252/45 LIEF020262/44 LIEF020262/43 LIEF020262/42 LIEF020262/41 LIEF020262/40 LIEF020262/39 LIEF020262/38 LIEF020262/37 LIEF020262/36 LIEF020262/46 LIEF020252/46 LIEF020262/47 LIEF020252/44 LIEF020252/43 LIEF020252/42 LIEF020252/41 LIEF020252/40 LIEF020252/39 LIEF020252/38 LIEF020252/37 LIEF020252/36
Occupational shoes, slip-on, Alpro®, A 640
Schoenen en laarzen
This occupational safety shoe is ideal for everyday work in catering or food industries. It comes with a replaceable, anatomically formed integrated insole made from cork.
  • Can be washed and disinfected at temperatures up to 60 °C
  • Integrated insole washable at 30 °C
Colour: Green, black, blue, white
Colour: Green, black, blue, white
Certificaten: EN ISO 20345:2011; SRC, SB E