Wash lotions, Baktolin® pure
Leverancier: HARTMANN PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
981328 981329
HART981328EA 73.1 EUR
HART981328 HART981329
Wash lotions, Baktolin® pure
Huidverzorging Zepen en huidreinigingsmiddelen
Baktolin® pure possesses a skin-neutral pH-value. Together with the skin-friendly surfactants it gently cleanses the skin and maintains its natural protective acid mantle. At the same time, this makes the product ideal for use in food-processing areas, in which any risk to the processed goods must be avoided.
  • Modern, wash-active substances and gentle preservatives ensure it is gentle on all skin types, including sensitive skin
  • Neutral pH value of 5,5
  • Free of alkali, soap, fragence or colourant
Opgelet: * These articles are only available in Germany.
*De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904