Overalls, Primeguard, PP
Leverancier: Synergy Health
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PG5XX11 PG5X11
VEROPG5XX11EA 323 EUR
VEROPG5XX11 VEROPG5X11
Overalls, Primeguard, PP
Wegwerpkleding
These disposable economy overalls are made of PP. They are designed for protection against airborne particles and limited non toxic splash and spray. Ideal for visitors.
  • Hood
  • Elasticated wrists and ankles
  • Anti-static
Colour: White or blue
Weight: 50 g/m²
Type 5/6