Neutral detergents for cleanrooms, ClearKlens Plus VH5
Leverancier: Diversey
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ClearKlens®
141-1377EA 74.7 EUR
141-1377 141-1378 141-1379
Neutral detergents for cleanrooms, ClearKlens Plus VH5
Reinigingsmiddelen Detergenten en oppervlaktereinigers
ClearKlens Plus contains a concentrated blend of detergents and emulsifiers. Its balanced neutral formulation is particularly effective against product residues found in the pharmaceutical and cosmetic industries.
  • Available in a 30 ml single-shot bottle of concentrate for ease of dilution
  • Compatible with materials commonly found in pharmaceutical cleanrooms
  • Single-shot bottle is 0,2 µm filtered, sterile filled and double-bagged
  • Supplied with Certificate of Analysis and sterility as appropriate