Cleanroom bond papers, Munising LP®
Leverancier: CONNECTICUT CLEANROOM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PB2S001BC PB2S002 PB3S001 PB3S002
CONNPB2S001BCEA 455 EUR
CONNPB2S001BC CONNPB2S002 CONNPB3S001 CONNPB3S002
Cleanroom bond papers, Munising LP®
Papier
Munising LP® bond papers are saturated with synthetic polymers and are free of inorganic fillers which prevents the doping of products and accepts standard printing inks without skipping, smudging or smearing.
  • Natural latex free
  • Heat resistant for use in laser printers and photocopy machines
  • Autoclavable