DuraStripe® X-Treme Floor Stripes
Leverancier: ERGOMAT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DSX3100Y DSX3100R DSX2100Y DSX2100B DSX2100R DSX2100W DSX2100G DSX4100Y DSX530Y DSX7.530R 4074300X5G 4076302X10B DSX4100BL DSX4100G DSX530DB 4074300X5NABLU 4074300X5WH 4074300X5RED 4074300X7.5WH
ERGODSX3100YEA 161 EUR
ERGODSX3100Y ERGODSX3100R ERGODSX2100Y ERGODSX2100B ERGODSX2100R ERGODSX2100W ERGODSX2100G ERGODSX4100Y ERGODSX530Y ERGODSX7.530R ERGO4074300X5G ERGO4076302X10B ERGODSX4100BL ERGODSX4100G ERGODSX530DB ERGO4074300X5NABLU ERGO4074300X5WH ERGO4074300X5RED ERGO4074300X7.5WH
DuraStripe® X-Treme Floor Stripes
Pictogrammen
DuraStripe® is the easiest, fastest and least expensive floor marking system for industrial floors. The durability and vast size of the DuraStripe® program makes it the only total floor marking solution available on the market today. If you find that painting aisle striping is too expensive and involves too much downtime, the DuraStripe program is the perfect solution for your facility floors.
  • Ideal for smooth or newer floors but can be used on old floors as well
  • Simple and easy peel-and-stick installation
  • Easy to remove for a quick reconfiguration and hence a great Lean manufacturing tool
  • No dry time, cure time, fumes or messy clean up to limit productivity
  • Available in 11 different colours
Made for critical environments and area where pivoting with forklifts/tow motors can occur. It has a special coated surface that facilitates easy cleaning.