Permanent marker pens, Pentel N50
Leverancier: DIALSTAT OFFICE SUPPLIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PEN50BU PEN50BK
DIALPEN50BUEA 0 EUR
DIALPEN50BU DIALPEN50BK
Permanent marker pens, Pentel N50
Pennen Permanente markeerstiften
Pentel N50 marker has bonded fibre tip which helps retain its shape. Will write on almost any surface, including cardboard, metal, wood, plastic and glass. The 4,3 mm tip gives ∼2,2 mm line width.
  • Robust aluminium barrel
  • Bullet point tip