High temperature anti-static cleanroom tapes
Leverancier: ULTRATAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2157BZ050-P3D 2157BZ075-P3D 2157BZ100-P3D 2157BZ150-P3D 2157BZ200-P3D 2157BZ025-P3D
819-0100EA 921 EUR
819-0100 819-0101 819-0102 819-0103 819-0104 UTAP2157BZ025-P3D
High temperature anti-static cleanroom tapes
Tape
High temperature polyimide ESD tapes with silicone adhesive. These cleanroom tapes are designed for use in ESD-sensitive environments such as sealing anti-static bags, packing of electronic components, medical instruments, precision mechanical equipment, and splicing films. They can withstand temperatures up to 315 °C for use during circuit board manufacturing.
  • Naturally anti-static or treated to produce an anti-static surface to generate very low voltage during unwind and use
Length: 32,9 m
Colour: Bronze