Cleaning swabs, MegaTip™
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QDMT-1 MT-6IR MT-6
129-1071EA 97.5 EUR
129-1071 129-1085 129-1070
Cleaning swabs, MegaTip™
Wattenstaafjes Wattenstaafjes voor reiniging
MegaTip are substantial cleaning swabs.
When used with the universal handle, it will easily reach to clean lighting recesses, corners and angles between walls and ceilings and other areas where a mop or wiping area is not ideal.
Certificaten: ISO 9001:2008.
Bestelinformatie: Handles and handle adapter need to be ordered separately.