Pressure loggers, C9500 series
Leverancier: COMARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C9551 C9553 C9555 C9555/SIL C9557
628-4204EA 1210 EUR
628-4204 628-4203 628-4202 COMAC9555/SIL 628-4201
Pressure loggers, C9500 series
Dataloggers
The C9500 pressure meters are designed for use with external pressure transducer. It measures the gauge/differential and positive/negative (vacuum) pressures.
  • Safe and acccurate
  • Suitable for dry or normal use
  • Measures the gauge or differential and ± pressures
It is applicable in the maintenance of plastic injection machines, hydraulic presses and other machinaries, mobile plants such as diggers, tractors and earth movers, production engineering (production and processing of food, pharmaceuticals, chemicals and petrochemicals), heating and refrigeration, gas production and distribution.
Lowest four digits blink over 9999 reading indication.
Certificaten: Approval by BASEEFA; accredited by UKAS calibration. IP 67, BS:EN60529, IEC529 protection.