Floor drain brush and handles, J-Zone
Leverancier: MICRONOVA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BRBL-46 BRHFD-36
129-0594EA 75.4 EUR
129-0594 129-0597
Floor drain brush and handles, J-Zone
Borstels
A stiff-bristle brush head and threaded plastic handle, specifically designed for cleaning floor drains. The brush head features black bristles for colour-coding and the handle is printed with 'For Floor Drain Use Only' to keep separate from other cleaning brushes.
  • Stiff bristle, replaceable brush head
  • Handle is printed with 'For Floor Drain Use Only'
  • Clear distinction from other cleaning brushes
  • 4" (10 cm) diameter brush head
  • Long, 36" (91 cm) handle to easily reach floor
The Micronova J-Zone Floor Drain Brush head features a plastic threaded end for easy replacement with the Floor Drain Handle.
Bestelinformatie: Brush and handle must be ordered separately.